Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
J3
Entrez Gene ID
823531
Gene Name
DNAJ homologue 3
Gene Aliases
AT3G44110, ATJ, ATJ3, DNAJ homologue 3
Chromosome Location
Chr.3: 15868631 - 15871256 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114279.4 NP_189997.1 1 - 2 345 90
NM_180322.2 NP_850653.1 1 - 2 345 90
GenBank mRNA AK226464.1 - 1 - 2 345 90
AY035087.1 - 1 - 2 264 90
AY088078.1 - 1 - 2 290 90
AY113878.1 - 1 - 2 148 90

View Full Product Details