Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VAT1
Entrez Gene ID
831490
Gene Name
VALINE-TOLERANT 1
Gene Aliases
AT5G16290, MQK4.1, MQK4_1, VALINE-TOLERANT 1
Chromosome Location
Chr.5: 5333750 - 5337784 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121634.4 NP_197133.1 4 - 5 711 118
NM_180498.3 NP_850829.1 4 - 5 698 118
GenBank mRNA AK226906.1 - 4 - 5 637 118
BT021989.1 - 4 - 5 600 118

View Full Product Details