Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDN2
Entrez Gene ID
830437
Gene Name
small RNA degrading nuclease 2
Gene Aliases
AT5G05540, MOP10.8, MOP10_8, small RNA degrading nuclease 2
Chromosome Location
Chr.5: 1636282 - 1638939 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125703.2 NP_001119175.1 6 - 7 764 130
NM_120636.4 NP_196173.1 6 - 7 764 130
GenBank mRNA AK118437.1 - 6 - 7 740 130
BT009685.1 - 6 - 7 627 130

View Full Product Details