Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDN2
Entrez Gene ID
830437
Gene Name
small RNA degrading nuclease 2
Gene Aliases
AT5G05540, MOP10.8, MOP10_8, small RNA degrading nuclease 2
Chromosome Location
Chr.5: 1636282 - 1638939 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125703.2 NP_001119175.1 8 - 9 943 93
NM_120636.4 NP_196173.1 8 - 9 943 93
GenBank mRNA AK118437.1 - 8 - 9 919 93
BT009685.1 - 8 - 9 806 93

View Full Product Details