Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UGT76C2
Entrez Gene ID
830471
Gene Name
UDP-glucosyl transferase 76C2
Gene Aliases
AT5G05860, MJJ3.28, MJJ3_28, UDP-glucosyl transferase 76C2
Chromosome Location
Chr.5: 1765390 - 1767510 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120668.4 NP_196205.1 1 - 2 632 80
GenBank mRNA AY045617.1 - 1 - 2 494 80
AY143896.1 - 1 - 2 477 80
KJ138703.1 - 1 - 2 477 80

View Full Product Details