Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MES8
- Entrez Gene ID
-
816891
- Gene Name
- methyl esterase 8
- Gene Aliases
-
AT2G23590, ATMES8, F26B6.24, F26B6_24, METHYL ESTERASE 8, methyl esterase 8
- Chromosome Location
-
Chr.2: 10034484 - 10036048 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127923.2 |
NP_179940.1 |
1 - 2 |
488 |
82 |
View Full Product Details