Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIMT1
Entrez Gene ID
823991
Gene Name
protein-l-isoaspartate methyltransferase 1
Gene Aliases
AT3G48330, ATPIMT1, Arabidopsis thaliana protein-l-isoaspartate methyltransferase 1, protein-l-isoaspartate methyltransferase 1
Chromosome Location
Chr.3: 17892738 - 17894162 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339361.1 NP_001326364.1 2 - 3 788 77
NM_001339362.1 NP_001326363.1 3 - 4 621 77
NM_148859.4 NP_680112.2 2 - 3 443 77
NM_180682.4 NP_851013.2 3 - 4 600 77
GenBank mRNA BT029372.1 - 2 - 3 443 77

View Full Product Details