Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AO1
Entrez Gene ID
832221
Gene Name
aldehyde oxidase 1
Gene Aliases
AAO1, ALDEHYDE OXIDASE, AOalpha, ARABIDOPSIS THALIANA ALDEHYDE OXIDASE 1, AT-AO1, AT5G20960, ATAO, Arabidopsis thaliana aldehyde oxidase 1, AtAO1, F22D1.130, F22D1_130, aldehyde oxidase 1, aldehyde oxidase alpha
Chromosome Location
Chr.5: 7116455 - 7122774 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122105.3 NP_568407.2 2 - 3 1931 80
NM_180718.2 NP_851049.1 2 - 3 2163 80
GenBank mRNA AB005804.1 - 2 - 3 1880 80

View Full Product Details