Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDAR1
Entrez Gene ID
824454
Gene Name
monodehydroascorbate reductase 1
Gene Aliases
AT3G52880, ATMDAR1, monodehydroascorbate reductase 1
Chromosome Location
Chr.3: 19601167 - 19604597 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115148.4 NP_190856.1 1 - 2 284 80
GenBank mRNA AF360197.1 - 1 - 2 79 80
AK227030.1 - 1 - 2 108 80
AK317341.1 - 1 - 2 107 80
AY045666.1 - 1 - 2 107 80
AY057576.1 - 1 - 2 135 80
AY060525.1 - 1 - 2 53 80
AY070718.1 - 1 - 2 126 80
AY091403.1 - 1 - 2 53 80
AY125492.1 - 1 - 2 105 80

View Full Product Details