Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EDA7
Entrez Gene ID
824866
Gene Name
embryo sac development arrest 7
Gene Aliases
AT3G56990, embryo sac development arrest 7
Chromosome Location
Chr.3: 21088027 - 21092169 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339835.1 NP_001327450.1 2 - 3 424 118
NM_001339836.1 NP_001327449.1 2 - 3 396 118
NM_115558.6 NP_191258.2 2 - 3 424 118
GenBank mRNA AY069919.1 - 2 - 3 311 118

View Full Product Details