Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HAF01
- Entrez Gene ID
-
840169
- Gene Name
- HAC13 protein (HAC13)
- Gene Aliases
-
AT1G32750, F6N18.13, F6N18_13, GTD01, GTD1, HAC13, HAF1, HISTONE ACETYLTRANSFERASE OF THE CBP FAMILY 13, HISTONE ACETYLTRANSFERASE OF THE CBP FAMILY 13, HISTONE ACETYLTRANSFERASE OF THE TAFII250 FAMILY 1, TAF1, TBP-ASSOCIATED FACTOR 1
- Chromosome Location
-
Chr.1: 11846247 - 11856765 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_103009.4 |
NP_174552.1 |
5 - 6 |
548 |
70 |
GenBank mRNA |
AF510669.1 |
- |
4 - 5 |
256 |
70 |
View Full Product Details