Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
840293
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AT1G33960, AVRRPT2-INDUCED GENE 1, F12G12.27
Chromosome Location
Chr.1: 12346138 - 12348733 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333075.1 NP_001322563.1 5 - 6 1023 98
NM_103118.4 NP_174658.1 5 - 6 929 98
GenBank mRNA AK176629.1 - 5 - 6 929 98
U40856.1 - 5 - 6 912 98

View Full Product Details