Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
840987
Gene Name
RAB GTPASE HOMOLOG B18
Gene Aliases
ARABIDOPSIS RAB GTPASE HOMOLOG B18, ARABIDOPSIS RAB GTPASE HOMOLOG C1, AT1G43890, ATRAB-C1, ATRAB18, ATRABC1, F28H19.15, F28H19_15, RAB GTPASE HOMOLOG 18-1, RAB GTPASE HOMOLOG B18, RAB GTPASE HOMOLOG C1, RAB18-1, RABC1
Chromosome Location
Chr.1: 16646696 - 16648648 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084206.1 NP_001077675.1 3 - 4 320 133
NM_001123963.1 NP_001117435.1 2 - 3 326 133
NM_103516.4 NP_175056.1 3 - 4 356 133
GenBank mRNA AF370263.1 - 3 - 4 280 133
AK227664.1 - 3 - 4 315 133
AY063080.1 - 2 - 3 209 133
AY086878.1 - 3 - 4 319 133
U75603.1 - 2 - 3 228 133

View Full Product Details