Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CER7
Entrez Gene ID
825221
Gene Name
3'-5'-exoribonuclease family protein
Gene Aliases
AT3G60500, ECERIFERUM 7, G3
Chromosome Location
Chr.3: 22353845 - 22356849 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203214.1 NP_001190143.1 9 - 10 1373 99
NM_001340047.1 NP_001327293.1 8 - 9 1498 99
NM_115914.4 NP_191609.1 9 - 10 1347 99
NM_202737.3 NP_974466.1 9 - 10 1419 99
GenBank mRNA BT004196.1 - 9 - 10 1300 99
BT005701.1 - 8 - 9 1127 99

View Full Product Details