Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDG9
Entrez Gene ID
831598
Gene Name
homeodomain GLABROUS 9
Gene Aliases
AT5G17320, MKP11.28, MKP11_28, homeodomain GLABROUS 9
Chromosome Location
Chr.5: 5703139 - 5707637 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121738.2 NP_197234.1 1 - 2 125 72

View Full Product Details