Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSD3
Entrez Gene ID
831928
Gene Name
copper/zinc superoxide dismutase 3
Gene Aliases
AT5G18100, COPPER/ZINC SUPEROXIDE DISMUTASE, MRG7.6, MRG7_6, copper/zinc superoxide dismutase 3
Chromosome Location
Chr.5: 5987187 - 5988885 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125773.1 NP_001119245.1 1 - 2 126 64
NM_121815.3 NP_197311.1 1 - 2 126 64
GenBank mRNA AK117742.1 - 1 - 2 115 64
BT003689.1 - 1 - 2 92 64

View Full Product Details