Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ORP1A
- Entrez Gene ID
-
817653
- Gene Name
- OSBP(oxysterol binding protein)-related protein 1A
- Gene Aliases
-
AT2G31020, F7F1.23, F7F1_23, OSBP(oxysterol binding protein)-related protein 1A
- Chromosome Location
-
Chr.2: 13201350 - 13204948 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128656.3 |
NP_180659.2 |
2 - 3 |
1145 |
104 |
View Full Product Details