Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPUORF7
Entrez Gene ID
817685
Gene Name
transcription factor bHLH155-like protein
Gene Aliases
AT2G31280, F16D14.12, F16D14_12, LHL2, LL2, LONESOME HIGHWAY LIKE 2, conserved peptide upstream open reading frame 7
Chromosome Location
Chr.2: 13338818 - 13343710 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036377.1 NP_001031454.1 9 - 10 2534 81
NM_128684.5 NP_180686.2 9 - 10 2803 81
GenBank mRNA AY954808.1 - 9 - 10 1943 81

View Full Product Details