Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MLO5
- Entrez Gene ID
-
817932
- Gene Name
- Seven transmembrane MLO family protein
- Gene Aliases
-
AT2G33670, ATMLO5, F4P9.44, F4P9_44, MILDEW RESISTANCE LOCUS O 5
- Chromosome Location
-
Chr.2: 14244951 - 14248155 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_128925.2 |
NP_180923.1 |
7 - 8 |
931 |
155 |
View Full Product Details