Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUP2
Entrez Gene ID
817941
Gene Name
purine permease 2
Gene Aliases
AT2G33750, ATPUP2, T1B8.6, T1B8_6, purine permease 2
Chromosome Location
Chr.2: 14271674 - 14273457 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_128934.3 NP_180931.2 1 - 2 923 125

View Full Product Details