Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAR3
Entrez Gene ID
841602
Gene Name
peptidase M20/M25/M40 family protein
Gene Aliases
AT1G51760, F19C24.4, F19C24_4, IAA-ALA HYDROLASE, IAA-ALANINE RESISTANT 3, JASMONIC ACID RESPONSIVE 3, JR3
Chromosome Location
Chr.1: 19199351 - 19201707 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104055.4 NP_175587.1 2 - 3 682 73
GenBank mRNA AF375444.1 - 2 - 3 565 73
AY143961.1 - 2 - 3 471 73

View Full Product Details