Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYSC1
Entrez Gene ID
825317
Gene Name
cysteine synthase C1
Gene Aliases
ARATH;BSAS3;1, AT3G61440, ATCYSC1, BETA-SUBSTITUTED ALA SYNTHASE 3;1, CYSTEINE SYNTHASE, CYSTEINE SYNTHASE C1, cysteine synthase C1
Chromosome Location
Chr.3: 22735663 - 22738131 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084855.2 NP_001078324.1 8 - 9 971 73
NM_001203223.1 NP_001190152.1 8 - 9 1173 73
NM_001340099.1 NP_001327883.1 8 - 9 1222 73
NM_116009.4 NP_191703.1 9 - 10 1215 73
GenBank mRNA AB024282.1 - 9 - 10 1063 73
AJ010505.1 - 9 - 10 1092 73
AK226606.1 - 9 - 10 1085 73
AY093094.1 - 9 - 10 1149 73
AY128782.1 - 9 - 10 1009 73

View Full Product Details