Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARF18
Entrez Gene ID
825356
Gene Name
auxin response factor 18
Gene Aliases
AT3G61830, F21F14.12, auxin response factor 18
Chromosome Location
Chr.3: 22887889 - 22891455 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340129.1 NP_001327870.1 9 - 10 1167 72
NM_116048.3 NP_567119.1 10 - 11 1275 72
GenBank mRNA AF334717.1 - 10 - 11 993 72
AY059746.1 - 10 - 11 1213 72
AY070463.1 - 10 - 11 1278 72
AY091392.1 - 10 - 11 993 72

View Full Product Details