Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF2
Entrez Gene ID
825828
Gene Name
protodermal factor 2
Gene Aliases
AT4G04890, T1J1.3, T1J1_3, protodermal factor 2
Chromosome Location
Chr.4: 2476390 - 2482447 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340517.1 NP_001329475.1 3 - 4 797 126
NM_116727.4 NP_567274.1 3 - 4 905 126
GenBank mRNA AF424560.1 - 3 - 4 767 126
AY062575.1 - 3 - 4 763 126
BT000144.1 - 2 - 3 355 126

View Full Product Details