Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATRPAC42
Entrez Gene ID
842377
Gene Name
DNA-directed RNA polymerase family protein
Gene Aliases
AAC42, AT1G60850, F23C21.3, F23C21_3, RNA POLYMERASE I SUBUNIT 42
Chromosome Location
Chr.1: 22397913 - 22400413 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333919.1 NP_001320360.1 2 - 3 586 83
NM_104766.5 NP_176282.1 3 - 4 480 83
NM_179502.4 NP_849833.1 3 - 4 469 83
NM_202328.3 NP_974057.1 2 - 3 485 83
GenBank mRNA AK226775.1 - 3 - 4 434 83

View Full Product Details