Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HT1
Entrez Gene ID
842538
Gene Name
Protein kinase superfamily protein
Gene Aliases
AT1G62400, F24O1.13, F24O1_13, high leaf temperature 1
Chromosome Location
Chr.1: 23089610 - 23091732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_104920.3 NP_176430.2 2 - 3 1400 58

View Full Product Details