Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MUB1
Entrez Gene ID
821307
Gene Name
membrane-anchored ubiquitin-fold protein 1 precursor
Gene Aliases
AT3G01050, T4P13.27, T4P13_27, membrane-anchored ubiquitin-fold protein 1 precursor
Chromosome Location
Chr.3: 13046 - 15906 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337306.1 NP_001319438.1 2 - 3 442 75
NM_001337307.1 NP_001327138.1 2 - 3 351 75
NM_001337308.1 NP_001327137.1 1 - 2 521 75
NM_001337309.1 NP_001327139.1 1 - 2 485 75
NM_110970.2 NP_186754.1 2 - 3 535 75
GenBank mRNA BT010661.1 - 2 - 3 319 75
BT012419.1 - 1 - 2 126 75

View Full Product Details