Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATCSLA15
Entrez Gene ID
826972
Gene Name
Nucleotide-diphospho-sugar transferases superfamily protein
Gene Aliases
AT4G13410, CELLULOSE SYNTHASE LIKE A15, CSLA15, T9E8.150, T9E8_150
Chromosome Location
Chr.4: 7792016 - 7796041 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001340860.1 NP_001329703.1 1 - 2 283 100
NM_001340861.1 NP_001329704.1 1 - 2 283 100
NM_117415.3 NP_193077.2 1 - 2 283 100
GenBank mRNA KJ138761.1 - 1 - 2 66 100

View Full Product Details