Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IAGLU
- Entrez Gene ID
-
827229
- Gene Name
- indole-3-acetate beta-D-glucosyltransferase
- Gene Aliases
-
AT4G15550, DL3815C, FCAALL.103, UDP-GLUCOSE:INDOLE-3-ACETATE BETA-D-GLUCOSYLTRANSFERASE, indole-3-acetate beta-D-glucosyltransferase
- Chromosome Location
-
Chr.4: 8877590 - 8879327 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117646.3 |
NP_567471.1 |
1 - 1 |
1541 |
141 |
View Full Product Details