Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRR2
Entrez Gene ID
827527
Gene Name
CheY-like two-component responsive regulator family protein
Gene Aliases
AT4G18020, PRR2, PSEUDO-RESPONSE REGULATOR 2, T6K21.200, T6K21_200
Chromosome Location
Chr.4: 10002846 - 10007626 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203830.1 NP_001190759.1 10 - 11 1733 98
NM_001341256.1 NP_001329341.1 10 - 11 1564 98
NM_117912.4 NP_567548.1 10 - 11 1746 98
NM_179072.3 NP_849403.1 10 - 11 1718 98
NM_179073.4 NP_849404.2 10 - 11 1686 98
GenBank mRNA AF385725.1 - 10 - 11 1570 98
AY143963.1 - 9 - 10 1426 98

View Full Product Details