Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APRR2
Entrez Gene ID
827527
Gene Name
CheY-like two-component responsive regulator family protein
Gene Aliases
AT4G18020, PRR2, PSEUDO-RESPONSE REGULATOR 2, T6K21.200, T6K21_200
Chromosome Location
Chr.4: 10002846 - 10007626 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084936.2 NP_001078405.1 4 - 5 629 85
NM_001160778.2 NP_001154250.1 4 - 5 665 85
NM_001203830.1 NP_001190759.1 4 - 5 652 85
NM_001341252.1 NP_001329342.1 5 - 6 643 85
NM_001341253.1 NP_001329343.1 5 - 6 623 85
NM_001341254.1 NP_001329339.1 4 - 5 904 85
NM_001341255.1 NP_001329340.1 4 - 5 884 85
NM_001341256.1 NP_001329341.1 4 - 5 483 85
NM_117912.4 NP_567548.1 4 - 5 665 85
NM_179072.3 NP_849403.1 4 - 5 637 85
NM_179073.4 NP_849404.2 4 - 5 605 85
GenBank mRNA AB041531.1 - 3 - 4 355 85
AF385725.1 - 4 - 5 489 85
AY143963.1 - 3 - 4 345 85

View Full Product Details