Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DOT4
- Entrez Gene ID
-
827609
- Gene Name
- Pentatricopeptide repeat (PPR) superfamily protein
- Gene Aliases
-
AT4G18750, DEFECTIVELY ORGANIZED TRIBUTARIES 4, F28A21.160, F28A21_160
- Chromosome Location
-
Chr.4: 10304662 - 10307673 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117991.3 |
NP_193610.1 |
1 - 1 |
2539 |
108 |
View Full Product Details