Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAO4
Entrez Gene ID
842894
Gene Name
polyamine oxidase 4
Gene Aliases
AT1G65840, ATPAO4, F1E22.18, F1E22_18, polyamine oxidase 4
Chromosome Location
Chr.1: 24489758 - 24493077 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105256.4 NP_176759.1 4 - 5 777 118
GenBank mRNA AF364953.1 - 4 - 5 731 118
BT000353.1 - 4 - 5 362 118

View Full Product Details