Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UPL2
Entrez Gene ID
843368
Gene Name
ubiquitin-protein ligase 2
Gene Aliases
AT1G70320, F17O7.15, F17O7_15, ubiquitin-protein ligase 2
Chromosome Location
Chr.1: 26488454 - 26501652 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_105700.3 NP_177189.1 1 - 2 407 84

View Full Product Details