Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APK1A
Entrez Gene ID
837271
Gene Name
Protein kinase superfamily protein
Gene Aliases
APK1, AT1G07570, F22G5.5, F22G5_5
Chromosome Location
Chr.1: 2331136 - 2333767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331708.1 NP_001318944.1 5 - 6 1020 98
NM_001331709.1 NP_001320609.1 4 - 5 977 98
NM_001331710.1 NP_001320608.1 5 - 6 926 98
NM_100631.4 NP_172237.1 4 - 5 945 98
NM_202049.3 NP_973778.1 5 - 6 974 98
GenBank mRNA BT004055.1 - 4 - 5 720 98
BT005112.1 - 4 - 5 640 98
D12522.1 - 4 - 5 717 98

View Full Product Details