Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIF3H1
Entrez Gene ID
837627
Gene Name
translation initiation factor 3 subunit H1
Gene Aliases
AT1G10840, T16B5.2, T16B5_2, translation initiation factor 3 subunit H1
Chromosome Location
Chr.1: 3607631 - 3610616 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100960.3 NP_563880.1 7 - 8 876 91
NM_202079.1 NP_973808.1 6 - 7 762 91
GenBank mRNA AY054641.1 - 7 - 8 710 91
AY081546.1 - 6 - 7 559 91
AY087338.1 - 7 - 8 778 91
BT000765.1 - 7 - 8 655 91

View Full Product Details