Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- FWA
- Entrez Gene ID
-
828658
- Gene Name
- FLOWERING WAGENINGEN
- Gene Aliases
-
AT4G25530, FLOWERING WAGENINGEN, HDG6, HOMEODOMAIN GLABROUS 6, M7J2.100, M7J2_100
- Chromosome Location
-
Chr.4: 13038360 - 13042464 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118685.3 |
NP_567722.1 |
4 - 5 |
472 |
141 |
View Full Product Details