Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPP1
Entrez Gene ID
828690
Gene Name
glycerol-3-phosphatase 1
Gene Aliases
AT4G25840, F14M19.120, F14M19_120, glycerol-3-phosphatase 1
Chromosome Location
Chr.4: 13138957 - 13141097 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118717.3 NP_567731.1 5 - 6 834 127

View Full Product Details