Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ASP5
Entrez Gene ID
829330
Gene Name
aspartate aminotransferase 5
Gene Aliases
AAT3, ASPARTATE AMINOTRANSFERASE, ASPARTATE AMINOTRANSFERASE DEFICIENT 3, AT4G31990, ATAAT1, F11C18.15, aspartate aminotransferase 5
Chromosome Location
Chr.4: 15470744 - 15473885 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036690.1 NP_001031767.1 4 - 5 517 94
NM_001203956.1 NP_001190885.1 4 - 5 474 94
NM_119351.3 NP_194927.1 3 - 4 536 94
NM_179152.3 NP_849483.1 4 - 5 549 94
GenBank mRNA AK316993.1 - 4 - 5 402 94
AK317084.1 - 3 - 4 537 94
AY054660.1 - 4 - 5 519 94
AY081506.1 - 3 - 4 309 94

View Full Product Details