The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001123759.1 | NP_001117231.1 | 3 - 4 | 460 | 80 |
NM_001197982.1 | NP_001184911.1 | 5 - 6 | 755 | 80 | |
NM_001331528.1 | NP_001322680.1 | 4 - 5 | 745 | 80 | |
NM_001331529.1 | NP_001322678.1 | 3 - 4 | 612 | 80 | |
NM_001331530.1 | NP_001322679.1 | 4 - 5 | 658 | 80 | |
NM_001331531.1 | NP_001322673.1 | 3 - 4 | 1832 | 80 | |
NM_001331532.1 | NP_001322674.1 | 4 - 5 | 1741 | 80 | |
NM_001331533.1 | NP_001322675.1 | 4 - 5 | 1714 | 80 | |
NM_001331534.1 | NP_001322676.1 | 2 - 3 | 1419 | 80 | |
NM_001331535.1 | NP_001322677.1 | 3 - 4 | 1328 | 80 | |
NM_100402.3 | NP_172015.1 | 3 - 4 | 1301 | 80 | |
NM_179265.2 | NP_849596.1 | 4 - 5 | 764 | 80 | |
GenBank mRNA | AK316776.1 | - | 4 - 5 | 669 | 80 |
AY037177.1 | - | 3 - 4 | 458 | 80 | |
AY091690.1 | - | 2 - 3 | 354 | 80 |