Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLX1
Entrez Gene ID
837731
Gene Name
glyoxalase/bleomycin resistance protein/dioxygenase superfamily protein
Gene Aliases
AT1G11840, ATGLX1, F12F1.32, F12F1_32, GLYOXALASE I, glyoxalase I homolog
Chromosome Location
Chr.1: 3995158 - 3997965 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035948.1 NP_001031025.1 8 - 9 897 94
NM_001035949.3 NP_001031026.1 8 - 9 1108 94
NM_001198039.1 NP_001184968.1 8 - 9 1030 94
NM_101055.4 NP_172648.1 8 - 9 1154 94
NM_179312.4 NP_849643.1 8 - 9 1129 94
NM_179313.2 NP_849644.2 8 - 9 874 94
GenBank mRNA AB050553.1 - 8 - 9 875 94
AF428272.1 - 8 - 9 875 94
AK316841.1 - 8 - 9 866 94
AK318730.2 - 8 - 9 870 94
AY074569.1 - 8 - 9 875 94
AY087874.1 - 8 - 9 910 94

View Full Product Details