Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDL2
Entrez Gene ID
820577
Gene Name
LSD1-like2
Gene Aliases
AT3G13682, LSD1-like2
Chromosome Location
Chr.3: 4478873 - 4481767 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112218.2 NP_187981.1 1 - 2 1788 76

View Full Product Details