Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP18
Entrez Gene ID
829295
Gene Name
ubiquitin-specific protease 18
Gene Aliases
AT4G31670, F28M20.140, F28M20_140, ubiquitin-specific protease 18
Chromosome Location
Chr.4: 15335743 - 15339613 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119316.5 NP_194895.1 10 - 11 1522 159
GenBank mRNA AY142532.1 - 10 - 11 1415 159

View Full Product Details