Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRO2
Entrez Gene ID
839411
Gene Name
ferric reduction oxidase 2
Gene Aliases
AT1G01580, ATFRO2, F22L4.12, F22L4_12, FERRIC CHELATE REDUCTASE DEFECTIVE 1, FERRIC REDUCTION OXIDASE 2, FRD1, ferric reduction oxidase 2
Chromosome Location
Chr.1: 208995 - 213082 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001331284.1 NP_001322968.1 5 - 6 1476 67
NM_100040.3 NP_171664.1 5 - 6 1263 67
GenBank mRNA AY302057.1 - 5 - 6 1076 67

View Full Product Details