Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAP14
Entrez Gene ID
819301
Gene Name
purple acid phosphatase 14
Gene Aliases
AT2G46880, ATPAP14, F14M4.29, purple acid phosphatase 14
Chromosome Location
Chr.2: 19264573 - 19266456 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130255.2 NP_182211.2 3 - 4 1194 119
NM_201975.3 NP_973704.1 2 - 3 1276 119
GenBank mRNA AY261790.1 - 3 - 4 1194 119
AY261791.1 - 2 - 3 1266 119

View Full Product Details