Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTK
Entrez Gene ID
841405
Gene Name
S-methyl-5-thioribose kinase
Gene Aliases
5-methylthioribose kinase 1, AT1G49820, ATMTK, F10F5.1, F10F5_1, MTK1, S-methyl-5-thioribose kinase
Chromosome Location
Chr.1: 18442882 - 18445350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103869.4 NP_564555.1 4 - 5 1006 69
GenBank mRNA AY057512.1 - 4 - 5 956 69
AY090315.1 - 4 - 5 883 69

View Full Product Details