Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THFS
Entrez Gene ID
841470
Gene Name
10-formyltetrahydrofolate synthetase
Gene Aliases
10-FORMYLTETRAHYDROFOLATE SYNTHETASE, 10-formyltetrahydrofolate synthetase, AT1G50480, F11F12.17
Chromosome Location
Chr.1: 18701882 - 18704938 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_103931.4 NP_564571.1 3 - 4 1664 111
GenBank mRNA AF162279.1 - 3 - 4 1485 111
AY054637.1 - 3 - 4 1583 111
AY081549.1 - 3 - 4 1482 111

View Full Product Details