Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPa6
Entrez Gene ID
830824
Gene Name
pyrophosphorylase 6
Gene Aliases
AT5G09650, AtPPa6, F17I14.160, F17I14_160, pyrophosphorylase 6
Chromosome Location
Chr.5: 2991149 - 2993213 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121002.3 NP_196527.1 1 - 2 360 100
GenBank mRNA AJ252210.1 - 1 - 2 266 100
AY045945.1 - 1 - 2 348 100
AY079355.1 - 1 - 2 264 100
AY551439.1 - 1 - 2 264 100

View Full Product Details