Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP60
Entrez Gene ID
821983
Gene Name
heat shock protein 60
Gene Aliases
AT3G23990, HEAT SHOCK PROTEIN 60-3B, HSP60-3B, heat shock protein 60
Chromosome Location
Chr.3: 8668772 - 8672578 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113304.5 NP_189041.1 10 - 11 1372 107
GenBank mRNA AY099594.1 - 10 - 11 1216 107
BT010398.1 - 10 - 11 1131 107

View Full Product Details