Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
APA1
Entrez Gene ID
837740
Gene Name
aspartic proteinase A1
Gene Aliases
AT1G11910, ATAPA1, F12F1.24, F12F1_24, aspartic proteinase A1
Chromosome Location
Chr.1: 4016712 - 4021043 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332010.1 NP_001322792.1 11 - 12 2029 84
NM_101062.4 NP_172655.1 11 - 12 1401 84
GenBank mRNA AY056387.1 - 11 - 12 1276 84
AY056403.1 - 11 - 12 1276 84
AY063974.1 - 11 - 12 1276 84
AY088657.1 - 11 - 12 1276 84
BT001980.1 - 10 - 11 1227 84

View Full Product Details